Product Details

SNP ID
rs56104583
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:155039516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTTTGTCCTGGGATACGCCTTG[A/G]CTGCCATCTATGTGGGTGAGTATGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCST1 PubMed Links

Gene Details

Gene
DCST1
Gene Name
DC-STAMP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143687.2 490 Missense Mutation ACT,GCT T101A NP_001137159.1
NM_152494.3 490 Missense Mutation ACT,GCT T126A NP_689707.2
Gene
DCST2
Gene Name
DC-STAMP domain containing 2
There are no transcripts associated with this gene.

Gene
LOC100505666
Gene Name
uncharacterized LOC100505666
There are no transcripts associated with this gene.

View Full Product Details