Product Details

SNP ID
rs55905370
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:205572757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTGTGCCAAAAGCCTACTGAGC[A/G]TCCAGTACCAGACATTGTCCTTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFSD4A PubMed Links

Gene Details

Gene
MFSD4A
Gene Name
major facilitator superfamily domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181644.4 Intron NP_857595.3
XM_011509229.2 Intron XP_011507531.1

View Full Product Details