Product Details

SNP ID
rs61812063
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:155277796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTCGGGTGTTCGGCAGCCACA[A/G]AGCAGTGGAAATCGAGCAGGAGCGG
Phenotype
MIM: 602989 MIM: 609973
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLK2 PubMed Links

Gene Details

Gene
CLK2
Gene Name
CDC like kinase 2
There are no transcripts associated with this gene.

Gene
HCN3
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020897.2 370 Missense Mutation AAA,AGA K69R NP_065948.1
XM_011509816.2 370 Intron XP_011508118.1
XM_011509817.2 370 Missense Mutation AAA,AGA K69R XP_011508119.1
XM_011509818.2 370 Intron XP_011508120.1
XM_017001918.1 370 Intron XP_016857407.1

View Full Product Details