Product Details

SNP ID
rs57296519
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:225488617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTCTCAAAATAGAATTCTTTCT[A/T]GCTCATTCAGTTTTGTTAGAAACAC
Phenotype
MIM: 609061
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ENAH PubMed Links

Gene Details

Gene
ENAH
Gene Name
enabled homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008493.1 11153 UTR 3 NP_001008493.1
NM_018212.4 11153 UTR 3 NP_060682.2
XM_005273187.2 11153 UTR 3 XP_005273244.1
XM_011544229.2 11153 Intron XP_011542531.2
XM_017001746.1 11153 UTR 3 XP_016857235.1
XM_017001747.1 11153 Intron XP_016857236.1
XM_017001748.1 11153 Intron XP_016857237.1
XM_017001749.1 11153 Intron XP_016857238.1
XM_017001750.1 11153 Intron XP_016857239.1
XM_017001751.1 11153 UTR 3 XP_016857240.1
XM_017001752.1 11153 Intron XP_016857241.1
XM_017001753.1 11153 UTR 3 XP_016857242.1
XM_017001754.1 11153 UTR 3 XP_016857243.1
XM_017001755.1 11153 UTR 3 XP_016857244.1
XM_017001756.1 11153 UTR 3 XP_016857245.1
XM_017001757.1 11153 UTR 3 XP_016857246.1

View Full Product Details