Product Details

SNP ID
rs61849056
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:225783334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTCAAATATAGGCATTCTGATG[A/G]GAACTTGTGTGTTAAAGTAACAGAT
Phenotype
MIM: 600707
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRP9 PubMed Links

Gene Details

Gene
SRP9
Gene Name
signal recognition particle 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130440.1 Intron NP_001123912.1
NM_003133.5 Intron NP_003124.1

View Full Product Details