Product Details

SNP ID
rs61786040
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:28679658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTAACTCTAGCTGATGTGAGGT[A/G]GAGACTCAATATAATATTTGTTGAA
Phenotype
MIM: 604409
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GMEB1 PubMed Links

Gene Details

Gene
GMEB1
Gene Name
glucocorticoid modulatory element binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319674.1 Intron NP_001306603.1
NM_006582.3 Intron NP_006573.2
NM_024482.2 Intron NP_077808.1
XM_011540518.2 Intron XP_011538820.1
XM_011540519.2 Intron XP_011538821.1
XM_011540521.2 Intron XP_011538823.1
XM_011540522.2 Intron XP_011538824.1
XM_017000087.1 Intron XP_016855576.1

View Full Product Details