Product Details

SNP ID
hCV88377021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:19255961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTCCCACAGCTTCGGAAATGTG[C/T]GGACACCTACAAGTACCTTTTCATC
Phenotype
MIM: 616846
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMC1 PubMed Links

Gene Details

Gene
EMC1
Gene Name
ER membrane protein complex subunit 1
There are no transcripts associated with this gene.

Gene
MRTO4
Gene Name
MRT4 homolog, ribosome maturation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016183.3 356 Missense Mutation GCG,GTG A34V NP_057267.2

View Full Product Details