Product Details

SNP ID
rs62541914
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:20112849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGACACGGGGGAGCTGGCAATT[G/T]TACCCTCCATGGTGTCACTCTCCTG
Phenotype
MIM: 605630
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G2D PubMed Links

Gene Details

Gene
PLA2G2D
Gene Name
phospholipase A2 group IID
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271814.1 1771 UTR 3 NP_001258743.1
NM_012400.3 1771 UTR 3 NP_036532.1

View Full Product Details