Product Details

SNP ID
rs62541892
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:20115512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAGGTCTGCGTACTCACAGCAG[C/T]GGATGTTCCCCTGGGAAAAGTTGTA
Phenotype
MIM: 605630
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G2D PubMed Links

Gene Details

Gene
PLA2G2D
Gene Name
phospholipase A2 group IID
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271814.1 355 Intron NP_001258743.1
NM_012400.3 355 Missense Mutation CAC,CGC H96R NP_036532.1

View Full Product Details