Product Details

SNP ID
rs56169842
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:25808728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGATCATCCCCAGTGAGCTGA[A/G]CATGTTCACTGGCTACCTGTCCAAC
Phenotype
MIM: 606210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPN1 PubMed Links

Gene Details

Gene
SEPN1
Gene Name
selenoprotein N, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020451.2 741 Missense Mutation AAC,AGC N229S NP_065184.2
NM_206926.1 741 Missense Mutation AAC,AGC N195S NP_996809.1

View Full Product Details