Product Details

SNP ID
rs57795102
Assay Type
Functionally Tested
NCBI dbSNP Submissions
24
Location
Chr.1:248361672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAATCCAAACATCCAATGGCCA[A/G]TATCACCTGGATGGCCAACCACACT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR2T4 PubMed Links

Gene Details

Gene
OR2T4
Gene Name
olfactory receptor family 2 subfamily T member 4
There are no transcripts associated with this gene.

View Full Product Details