Product Details

SNP ID
rs55871516
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:248681658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCACCGTTGAGAAGATTTGGAAA[C/T]AGGAGATCATCATGAGAATAAAGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR14I1 PubMed Links

Gene Details

Gene
OR14I1
Gene Name
olfactory receptor family 14 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004734.1 782 Missense Mutation TAT,TGT Y216C NP_001004734.1
XM_005273132.3 782 Missense Mutation TAT,TGT Y216C XP_005273189.1

View Full Product Details