Product Details

SNP ID
rs55957512
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:13478283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGCACTTACTATGTGCAGATCTT[G/T]TTCTAATCACTACACACGTTTTTAC
Phenotype
MIM: 615212
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRRC38 PubMed Links

Gene Details

Gene
LRRC38
Gene Name
leucine rich repeat containing 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010847.1 Intron NP_001010847.1

View Full Product Details