Product Details

SNP ID
rs56131445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:237863540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTCCCAGCCCAAACAAAATCG[C/T]ACTCACTTCACCGAATGATGTTATT
Phenotype
MIM: 605153
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAMP1 PubMed Links

Gene Details

Gene
RAMP1
Gene Name
receptor activity modifying protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308353.1 Intron NP_001295282.1
NM_005855.3 Intron NP_005846.1
XM_017003152.1 Intron XP_016858641.1
XM_017003153.1 Intron XP_016858642.1
XM_017003154.1 Intron XP_016858643.1
XM_017003155.1 Intron XP_016858644.1
XM_017003156.1 Intron XP_016858645.1

View Full Product Details