Product Details
- SNP ID
-
hCV88423459
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:170818597 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCCGGGACGACCGAGCTGATGGC[A/G]TCTTCGACCCCATCTTCGTCCGCAA
- Phenotype
-
MIM: 605363
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAD1
PubMed Links
Gene Details
- Gene
- GAD1
- Gene Name
- glutamate decarboxylase 1
There are no transcripts associated with this gene.
View Full Product Details