Product Details

SNP ID
rs62167156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151271278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGAGGAAAAGGGAGCAGGTCT[C/G]GGGTAAAGATAATGAGTTCAGAGAA
Phenotype
MIM: 603525
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NMI PubMed Links

Gene Details

Gene
NMI
Gene Name
N-myc and STAT interactor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004688.2 Intron NP_004679.2
XM_005246941.2 Intron XP_005246998.1
XM_017005247.1 Intron XP_016860736.1
Gene
RBM43
Gene Name
RNA binding motif protein 43
There are no transcripts associated with this gene.

View Full Product Details