Product Details
- SNP ID
-
hCV88455485
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:216113126 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTGATAACCATGTTTGTACAGCG[A/G]CAGGTAAGTTTCAGATTGACACTGA
- Phenotype
-
MIM: 194364
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
XRCC5
PubMed Links
Gene Details
- Gene
- XRCC5
- Gene Name
- X-ray repair cross complementing 5
There are no transcripts associated with this gene.
View Full Product Details