Product Details
- SNP ID
-
hCV88455891
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:216501350 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATCTTTCTTTCCAGTACCACTTC[A/C]GCTGTCACGGTAAAGTCCGCCATCA
- Phenotype
-
MIM: 613314
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RPL37A
PubMed Links
Gene Details
- Gene
- RPL37A
- Gene Name
- ribosomal protein L37a
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000998.4 |
|
Intron |
|
|
NP_000989.1 |
View Full Product Details