Product Details
- SNP ID
-
rs62191854
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:219289948 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAGAGCCAGGCCAGGGAATGTAG[G/T]CAGGAGAAGGCTCTGGGTAGAATTG
- Phenotype
-
MIM: 604139
MIM: 601773
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAJB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148769551,rs563465552] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAJB2
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B2
There are no transcripts associated with this gene.
- Gene
- MIR153-1
- Gene Name
- microRNA 153-1
There are no transcripts associated with this gene.
- Gene
- PTPRN
- Gene Name
- protein tyrosine phosphatase, receptor type N
View Full Product Details