Product Details

SNP ID
rs62187223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222866657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTAACCTTTGAAGGCATGATTGT[A/G]TAGGGAAAGGAGCTGTGATGACATT
Phenotype
MIM: 602371
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSL3 PubMed Links

Gene Details

Gene
ACSL3
Gene Name
acyl-CoA synthetase long-chain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004457.3 Intron NP_004448.2
NM_203372.1 Intron NP_976251.1
XM_017003584.1 Intron XP_016859073.1

View Full Product Details