Product Details

SNP ID
rs62194575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233060576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGGGAGCTTCCTCGTGCGTGCCA[A/G]CGAGTCCATCTCCCGGGCATACGCG
Phenotype
MIM: 601582
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INPP5D PubMed Links

Gene Details

Gene
INPP5D
Gene Name
inositol polyphosphate-5-phosphatase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017915.2 610 Missense Mutation AAC,AGC N33S NP_001017915.1
NM_005541.4 610 Missense Mutation AAC,AGC N33S NP_005532.2
XM_017004004.1 610 UTR 5 XP_016859493.1

View Full Product Details