Product Details

SNP ID
rs62186361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241229103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCTTCCTTGCACCCTGACACCT[A/G]TGCTGAGGCGCTCTGGCAGCTGGAG
Phenotype
MIM: 605096 MIM: 142695
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO7 PubMed Links

Gene Details

Gene
ANO7
Gene Name
anoctamin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001666.3 4533 Intron NP_001001666.1
NM_001001891.3 4533 Intron NP_001001891.2
XM_011511263.2 4533 Intron XP_011509565.1
XM_011511267.2 4533 Intron XP_011509569.1
XM_017004229.1 4533 Intron XP_016859718.1
XM_017004230.1 4533 Intron XP_016859719.1
Gene
HDLBP
Gene Name
high density lipoprotein binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243900.2 4533 UTR 3 NP_001230829.1
NM_001320965.1 4533 UTR 3 NP_001307894.1
NM_001320966.1 4533 UTR 3 NP_001307895.1
NM_001320967.1 4533 Intron NP_001307896.1
NM_005336.5 4533 UTR 3 NP_005327.1
NM_203346.4 4533 UTR 3 NP_976221.1
XM_005247002.3 4533 UTR 3 XP_005247059.2
XM_005247003.4 4533 UTR 3 XP_005247060.2
XM_006712475.3 4533 UTR 3 XP_006712538.1
XM_011511058.2 4533 UTR 3 XP_011509360.1
XM_011511060.2 4533 UTR 3 XP_011509362.1
XM_017003940.1 4533 UTR 3 XP_016859429.1
XM_017003941.1 4533 Intron XP_016859430.1
XM_017003942.1 4533 Intron XP_016859431.1

View Full Product Details