Product Details

SNP ID
rs59022946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:109294053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTAAGAGCCGCTTTGAGCTATA[C/T]GCAGAGTTTGGGGTCAAGAGAGTTA
Phenotype
MIM: 614445
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPPA2 PubMed Links

Gene Details

Gene
DPPA2
Gene Name
developmental pluripotency associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138815.3 Intron NP_620170.3
XM_011512443.1 Intron XP_011510745.1
XM_011512444.2 Intron XP_011510746.1
XM_011512445.2 Intron XP_011510747.1

View Full Product Details