Product Details

SNP ID
rs58879727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122565028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTGTGTTTTTGAGGATTACAAA[A/G]GTGATATATGCTCACTATAGAAATA
Phenotype
MIM: 613143 MIM: 612065
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DTX3L PubMed Links

Gene Details

Gene
DTX3L
Gene Name
deltex E3 ubiquitin ligase 3L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138287.3 Intron NP_612144.1
Gene
PARP9
Gene Name
poly(ADP-ribose) polymerase family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146102.1 Intron NP_001139574.1
NM_001146103.1 Intron NP_001139575.1
NM_001146104.1 Intron NP_001139576.1
NM_001146105.1 Intron NP_001139577.1
NM_001146106.1 Intron NP_001139578.1
NM_031458.2 Intron NP_113646.2
XM_005247820.2 Intron XP_005247877.1
XM_011513218.1 Intron XP_011511520.1
XM_017007304.1 Intron XP_016862793.1
XM_017007305.1 Intron XP_016862794.1
XM_017007306.1 Intron XP_016862795.1

View Full Product Details