Product Details

SNP ID
rs61761685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:124763314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCATTCCTGGAAGGGAGAGAC[A/G]GCCCAGCATCTCAGTATTTCATTGG
Phenotype
MIM: 147561
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGB5 PubMed Links

Gene Details

Gene
ITGB5
Gene Name
integrin subunit beta 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002213.4 2785 UTR 3 NP_002204.2
XM_005247436.2 2785 UTR 3 XP_005247493.1
XM_006713630.2 2785 UTR 3 XP_006713693.1
XM_017006351.1 2785 UTR 3 XP_016861840.1
XM_017006352.1 2785 UTR 3 XP_016861841.1
XM_017006353.1 2785 UTR 3 XP_016861842.1
XM_017006354.1 2785 UTR 3 XP_016861843.1
XM_017006355.1 2785 UTR 3 XP_016861844.1

View Full Product Details