Product Details
- SNP ID
-
rs62284032
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:150618314 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGTGGATGGCTATTGTGAATCAAA[C/T]GAGAGAGTGAATTGGAGAGATTGTG
- Phenotype
-
MIM: 607912
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SELT
PubMed Links
Gene Details
- Gene
- SELT
- Gene Name
- selenoprotein T
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016275.3 |
|
Intron |
|
|
NP_057359.2 |
View Full Product Details