Product Details

SNP ID
rs56084310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:165773369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGGAAAATATGTTCTATAAAGG[A/G]TAAATCTATTAATTAGAGACCCACA
Phenotype
MIM: 177400
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCHE PubMed Links

Gene Details

Gene
BCHE
Gene Name
butyrylcholinesterase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017006967.1 1984 UTR 3 XP_016862456.1

View Full Product Details