Product Details
- SNP ID
-
rs56204521
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:170996983 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTGATCAATCCTTTACTTCAAATT[G/T]TCTTACATTAGATTTGATCAAACTT
- Phenotype
-
MIM: 138160
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC2A2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77733690] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC2A2
- Gene Name
- solute carrier family 2 member 2
View Full Product Details