Product Details

SNP ID
rs60191176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179327572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTACTTACAGGCATTTTTTAC[C/T]GTCTACAGAAACTTATTGTAATTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF639 PubMed Links

Gene Details

Gene
ZNF639
Gene Name
zinc finger protein 639
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303425.1 675 UTR 5 NP_001290354.1
NM_001303426.1 675 UTR 5 NP_001290355.1
NM_016331.2 675 UTR 5 NP_057415.1
XM_017006550.1 675 UTR 5 XP_016862039.1
XM_017006551.1 675 UTR 5 XP_016862040.1
XM_017006552.1 675 UTR 5 XP_016862041.1
XM_017006553.1 675 UTR 5 XP_016862042.1

View Full Product Details