Product Details
- SNP ID
-
rs61158071
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:179396766 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTAATTAATAAGTTACTTTTTAA[A/T]ATAAAGTGCTAGCTTAGATATTCTA
- Phenotype
-
MIM: 610863
MIM: 608506
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GNB4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs67376707] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GNB4
- Gene Name
- G protein subunit beta 4
- Gene
- MFN1
- Gene Name
- mitofusin 1
There are no transcripts associated with this gene.
View Full Product Details