Product Details

SNP ID
rs62295812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:192141836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATCATCACTACAAAAGCTCAAG[C/T]CATTTCATGCTAAAAGAGCCAGTGT
Phenotype
MIM: 601513
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF12 PubMed Links

Gene Details

Gene
FGF12
Gene Name
fibroblast growth factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004113.5 2816 UTR 3 NP_004104.3
NM_021032.4 2816 UTR 3 NP_066360.1
XM_005247227.2 2816 UTR 3 XP_005247284.1
XM_006713538.3 2816 UTR 3 XP_006713601.1
XM_017005879.1 2816 UTR 3 XP_016861368.1

View Full Product Details