Product Details

SNP ID
rs57769362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6708264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTAGTGTCTTCCATTGCACCTCT[A/G]AAGTTGGAAGATTCTGATCCCACGT
Phenotype
MIM: 605695
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BLOC1S4 PubMed Links

Gene Details

Gene
BLOC1S4
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 4
There are no transcripts associated with this gene.

Gene
MRFAP1L1
Gene Name
Morf4 family associated protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203462.2 1029 UTR 3 NP_982287.1
XM_005247932.1 1029 UTR 3 XP_005247989.1

View Full Product Details