Product Details

SNP ID
rs61184934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70711577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCAACCCTGTTAAGAAGGCA[A/G]AGAGCCATTTTCTGATCCATTAAGT
Phenotype
MIM: 611194
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RUFY3 PubMed Links

Gene Details

Gene
RUFY3
Gene Name
RUN and FYVE domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037442.3 Intron NP_001032519.1
NM_001130709.1 Intron NP_001124181.1
NM_001291993.1 Intron NP_001278922.1
NM_001291994.1 Intron NP_001278923.1
NM_014961.4 Intron NP_055776.1
XM_005265656.2 Intron XP_005265713.1
XM_005265658.1 Intron XP_005265715.1
XM_011531750.2 Intron XP_011530052.1
XM_011531751.2 Intron XP_011530053.1
XM_011531752.2 Intron XP_011530054.1
XM_011531753.2 Intron XP_011530055.1
XM_011531754.2 Intron XP_011530056.1
XM_011531755.2 Intron XP_011530057.1
XM_011531756.2 Intron XP_011530058.1
XM_017007890.1 Intron XP_016863379.1
XM_017007891.1 Intron XP_016863380.1
XM_017007892.1 Intron XP_016863381.1

View Full Product Details