Product Details

SNP ID
rs55947729
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:40336207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCTTAGATAATCTAAAGAAAGA[C/T]GGGGTTACATACTGTATGCAGCCAT
Phenotype
MIM: 605116
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CHRNA9 PubMed Links

Gene Details

Gene
CHRNA9
Gene Name
cholinergic receptor nicotinic alpha 9 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017581.3 Intron NP_060051.2
Gene
LOC101060498
Gene Name
uncharacterized LOC101060498
There are no transcripts associated with this gene.

View Full Product Details