Product Details

SNP ID
rs62303696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:46925033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTAAGAACAGGAATGGGGAGAG[C/T]GGAGATATTTTTCTAGGCCAGGAAG
Phenotype
MIM: 137141
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABRA4 PubMed Links

Gene Details

Gene
GABRA4
Gene Name
gamma-aminobutyric acid type A receptor alpha4 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000809.3 4717 UTR 3 NP_000800.2
NM_001204266.1 4717 UTR 3 NP_001191195.1
NM_001204267.1 4717 UTR 3 NP_001191196.1

View Full Product Details