Product Details

SNP ID
rs56264807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1463474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTCGCACAGTAAGCGTCGGTTC[C/T]GCAACACACTTCACAAAGGAACAAG
Phenotype
MIM: 610472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LPCAT1 PubMed Links

Gene Details

Gene
LPCAT1
Gene Name
lysophosphatidylcholine acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005248373.3 1210 UTR 3 XP_005248430.1
XM_011514132.1 1210 UTR 3 XP_011512434.1
XM_011514134.1 1210 UTR 3 XP_011512436.1
XM_017009879.1 1210 UTR 3 XP_016865368.1

View Full Product Details