Product Details

SNP ID
rs55686708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:32355828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAATCCATCAACTCCATCACTAT[C/T]TCTTCTTCGTTTCCTGTTGTTGTGG
Phenotype
MIM: 615635
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFR PubMed Links

Gene Details

Gene
ZFR
Gene Name
zinc finger RNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016107.3 3237 Missense Mutation AAT,GAT N1053D NP_057191.2

View Full Product Details