Product Details

SNP ID
rs56814521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:60597072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGAATTTATCCAGTTCAACTTT[C/G]CATTTTTAGCTAAGACTGAAATTAT
Phenotype
MIM: 616073
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DEPDC1B PubMed Links

Gene Details

Gene
DEPDC1B
Gene Name
DEP domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145208.1 2337 UTR 3 NP_001138680.1
NM_018369.2 2337 UTR 3 NP_060839.2
XM_011543509.2 2337 UTR 3 XP_011541811.1
XM_011543510.2 2337 Intron XP_011541812.1
XM_017009629.1 2337 Intron XP_016865118.1

View Full Product Details