Product Details

SNP ID
rs55759439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:64511196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGGAGAAGAAAACTCAGACTAT[G/T]TTTTTTCTAACTGGCCCTGTTTAAG
Phenotype
MIM: 610890
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RGS7BP PubMed Links

Gene Details

Gene
RGS7BP
Gene Name
regulator of G-protein signaling 7 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029875.2 Intron NP_001025046.1
NM_001271890.1 Intron NP_001258819.1
NM_001271891.1 Intron NP_001258820.1
XM_005248502.3 Intron XP_005248559.1

View Full Product Details