Product Details

SNP ID
rs56926064
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:170580060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGAAAAATTTTATCTTCCCTG[G/T]ATTCATGTTTTGCCATGGAATCCAG
Phenotype
MIM: 600866 MIM: 600075
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
PDCD2 PubMed Links

Gene Details

Gene
PDCD2
Gene Name
programmed cell death 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199461.1 815 Intron NP_001186390.1
NM_001199462.1 815 Missense Mutation NP_001186391.1
NM_001199463.1 815 Intron NP_001186392.1
NM_001199464.1 815 Intron NP_001186393.1
NM_002598.3 815 Missense Mutation NP_002589.2
NM_144781.2 815 Intron NP_659005.1
XM_017010926.1 815 Missense Mutation XP_016866415.1
Gene
TBP
Gene Name
TATA-box binding protein
There are no transcripts associated with this gene.

View Full Product Details