Product Details

SNP ID
hCV88748172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24429116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCCAGCAGCAATGACGACTTG[A/G]TTCTGTAAGGGACACAAGACAGAAT
Phenotype
MIM: 602515
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPLD1 PubMed Links

Gene Details

Gene
GPLD1
Gene Name
glycosylphosphatidylinositol specific phospholipase D1
There are no transcripts associated with this gene.

Gene
MRS2
Gene Name
MRS2, magnesium transporter
There are no transcripts associated with this gene.

View Full Product Details