Product Details

SNP ID
rs55711161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27957477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTTACACCACATGTACAGTCCC[A/T]CAAATGCTAGTAAATTTATGCAGCA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2B6 PubMed Links

Gene Details

Gene
OR2B6
Gene Name
olfactory receptor family 2 subfamily B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012367.1 237 Silent Mutation CCA,CCT P79P NP_036499.1

View Full Product Details