Product Details

SNP ID
rs61328033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52676090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCAGTGAAGAGACCTGGCTGAC[C/T]AGAGCAGAAGTTTTATTTGGGTAAG
Phenotype
MIM: 616870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC730101 PubMed Links

Gene Details

Gene
LOC730101
Gene Name
uncharacterized LOC730101
There are no transcripts associated with this gene.

Gene
TMEM14A
Gene Name
transmembrane protein 14A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014051.3 Intron NP_054770.1

View Full Product Details