Product Details

SNP ID
rs59781194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:53272577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTAGTATTAATTAGTTCATAGA[C/T]GAATTCATGAGAGGAGAGAAAGAGG
Phenotype
MIM: 611805 MIM: 603715
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELOVL5 PubMed Links

Gene Details

Gene
ELOVL5
Gene Name
ELOVL fatty acid elongase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242828.1 Intron NP_001229757.1
NM_001242830.1 Intron NP_001229759.1
NM_001242831.1 Intron NP_001229760.1
NM_001301856.1 Intron NP_001288785.1
NM_021814.4 Intron NP_068586.1
Gene
GCM1
Gene Name
glial cells missing homolog 1
There are no transcripts associated with this gene.

Gene
MIR5685
Gene Name
microRNA 5685
There are no transcripts associated with this gene.

View Full Product Details