Product Details

SNP ID
rs62435468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1671958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATACGGTGGTCAGGGAGTTCTGA[G/T]CCCCACCCCTAGTGGGCAGAAGACC
Phenotype
MIM: 614964
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ELFN1 PubMed Links

Gene Details

Gene
ELFN1
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128636.2 1414 Intron NP_001122108.1
XM_006715725.3 1414 Intron XP_006715788.1
XM_006715726.2 1414 Intron XP_006715789.1
XM_006715727.3 1414 Intron XP_006715790.1
XM_011515397.2 1414 Intron XP_011513699.1
XM_011515398.2 1414 Intron XP_011513700.1
XM_011515401.2 1414 Intron XP_011513703.1
XM_017012203.1 1414 UTR 5 XP_016867692.1

View Full Product Details