Product Details
- SNP ID
-
rs58370921
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:28956535 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGCAGGTCTAGGGACTGTGGAGC[C/T]GGGCCCGCGGCGGCAGCGACGGAGG
- Phenotype
-
MIM: 613356
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TRIL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1006521] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRIL
- Gene Name
- TLR4 interactor with leucine rich repeats
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014817.3 |
1879 |
Silent Mutation |
CCA,CCG |
P504P |
NP_055632.2 |
View Full Product Details