Product Details

SNP ID
rs55835817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44206115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGGCCATGTTGTCAAGGGTGT[A/G]TTTTCTGCACCACTGGCTGTTTCTT
Phenotype
MIM: 606209
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YKT6 PubMed Links

Gene Details

Gene
YKT6
Gene Name
YKT6 v-SNARE homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006555.3 Intron NP_006546.1
XM_005249582.4 Intron XP_005249639.1
XM_017011682.1 Intron XP_016867171.1

View Full Product Details