Product Details

SNP ID
rs60923628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:45724316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTACTGATGACAAATGTTCATC[A/G]CATCTAAAAGATACCTTCAAAGCAA
Phenotype
MIM: 103072 MIM: 611563
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY1 PubMed Links

Gene Details

Gene
ADCY1
Gene Name
adenylate cyclase 1
There are no transcripts associated with this gene.

Gene
SEPT7P2
Gene Name
septin 7 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details