Product Details

SNP ID
rs62441138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:5297342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGTGGAGACTCCTTCCTGCCAG[C/T]CTCCTTTATTCTGTGTACTCTTACT
Phenotype
MIM: 609149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC29A4 PubMed Links

Gene Details

Gene
SLC29A4
Gene Name
solute carrier family 29 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040661.1 Intron NP_001035751.1
NM_001300847.1 Intron NP_001287776.1
NM_153247.2 Intron NP_694979.2
XM_005249658.4 Intron XP_005249715.1
XM_006715667.3 Intron XP_006715730.1
XM_006715668.2 Intron XP_006715731.1
XM_011515200.2 Intron XP_011513502.1
XM_011515201.2 Intron XP_011513503.1
Gene
TNRC18
Gene Name
trinucleotide repeat containing 18
There are no transcripts associated with this gene.

View Full Product Details