Product Details

SNP ID
rs61731558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:5593428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGCGGCCGGCCGACGAGATCGC[A/C]GTGGACCGCGACGTGCCCTGGGGCG
Phenotype
MIM: 602689
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FSCN1 PubMed Links

Gene Details

Gene
FSCN1
Gene Name
fascin actin-bundling protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003088.3 Intron NP_003079.1

View Full Product Details